Bly(II)nitrat 99.999%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Lead dinitrate
193320100 193320500
ACRO193320100EA 1500 SEK
ACRO193320100 ACRO193320500
Bly(II)nitrat 99.999%
Bly(II)nitrat
Formel: Pb(NO₃)₂
Smältpunkt: 470 °C (dec.)
Densitet: 4,53 g/cm³
Storage Temperature: Omgivande
MDL Number: MFCD00011153
CAS nummer: 10099-74-8
EINECS: 233-245-9
UN: 1469
ADR: 5.1,II
Merck Index: 13,05432
Order Now
Specifikation testresultat
Appearance White Crystalline solid
Total trace metal impurities ≤0.001 %