Bly(II)klorid, ultra dry ≥99.999%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Lead dichloride
Ampuled under argon
42841.04 42841.09 42841.18
42841.04EA 1675 SEK
42841.04 42841.09 42841.18
Bly(II)klorid, ultra dry ≥99.999%
Bly(II)klorid
Purity: 99.999% CAS No.: 7758-95-4 Molecular Formula: PbCl2 Formula Weight: 278.10 Physical Form: Powder, Ampouled under argon Density: 5.85 Melting Point: 501° Boiling Point: 951° Merck Reference: 14,5404 UN No.: 2291 MDL No.: MFCD00011157
Formel: Cl₂Pb
Kokpunkt: 951 °C
Smältpunkt: 501 °C
Densitet: 5,85
Storage Temperature: Omgivande
MDL Number: MFCD00011157
CAS nummer: 7758-95-4
EINECS: 231-845-5
UN: 2291
ADR: 6.1,III
Merck Index: 14,5404