Kiselkarbid, grovt pulver 46 grit
Leverantör: Thermo Fisher Scientific
Varning
A14470.0C A14470.0E A14470.36
A14470.0CEA 13475 SEK
A14470.0C A14470.0E A14470.36
Kiselkarbid, grovt pulver 46 grit
Kiselkarbid
CAS No.: 409-21-2 Molecular Formula: SiC Formula Weight: 40.10 Odour: Odourless MDL No.: MFCD00049531
Formel: SiC
MW: 40.1 g/mol
Smältpunkt: 2700
Densitet: 3.16
Storage Temperature: Omgivande
MDL Number: MFCD00049531
CAS nummer: 409-21-2
EINECS: 206-991-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0bd532b391b9c28edcb8adac5fe5ed958ccaa487&vid=d39ce1cfd6c1a5ca44c7d11b656758cbfaed190d&prid=web&clid=SAPDAM