Aluminiumsulfatoktadekahydrat, extra pure
Leverantör: Thermo Scientific
Fara
Synonyms: Aluminum(III) sulphate octadecahydrate
123680010 123680025 123680100
ACRO123680010EA 1200 SEK
ACRO123680010 ACRO123680025 ACRO123680100
Aluminiumsulfatoktadekahydrat, extra pure
Aluminiumsulfatoktadekahydrat
Formel: Al₂(SO₄)₃·18H₂O
MW: 666,43 g/mol
Smältpunkt: 86 °C (dec.)
Densitet: 1,69 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00149136
CAS nummer: 7784-31-8
EINECS: 233-135-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4dace68759944aa1c6c0f56a372ea396f004e261&vid=e63de3c58650a97622a605c237aa52402ff157e4&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White Crystalline powder or crystals and/or chunks
Titration Complexometric 98.0 to 110.0 %
Chloride (Cl) ≤50 ppm
Iron (Fe) ≤50 ppm
Heavy metals (as Pb) ≤20 ppm
Insoluble matter ≤0.01 %