Aluminium sulphate 99.999%, extra pure
Leverantör: Thermo Fisher Scientific
Fara
192430050 192430250 192431000
ACRO192430050EA 935 SEK
ACRO192430050 ACRO192430250 ACRO192431000
Aluminium sulphate 99.999%, extra pure
Aluminium sulphate
Formel: Al₂O₁₂S₃
Smältpunkt: 770 °C
Densitet: 2,672 g/cm³ (22.5 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003423
CAS nummer: 10043-01-3
EINECS: 233-135-0
Merck Index: 12,00381
Order Now
Specifikation testresultat
Appearance White Powder and/or chunks
Titration Complexometric ≥15 % (Al) Typical
Total trace metal impurities ≤0.001 %