Aluminiumoxid, Supelco®
Leverantör: Merck
Fara
Synonyms: Aluminium(III) oxide, Alumina C
M-Clarity™ quality level = MQ200
1.15754.0100 1.15754.1000
1.15754.0100EA 1480 SEK
1.15754.0100 1.15754.1000
Aluminiumoxid, Supelco®
Aluminium oxide
Formel: Al₂O₃
MW: 101,96 g/mol
Kokpunkt: 2980 °C (1013 hPa)
Smältpunkt: 2045 °C
Densitet: 3,94 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003424
CAS nummer: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 13,00355