Aluminiumoxid, single crystal
Leverantör: Thermo Scientific
Synonyms: Aluminium(III) oxide, Alumina C
40261.KG 40261.KJ
40261.KGEA 1500 SEK
40261.KG 40261.KJ
Aluminiumoxid, single crystal
Aluminium oxide
CAS No.: 1344-28-1 Molecular Formula: Al2O3 Physical Form: 12.7mm dia x 1.0mm thick, random orientation MDL No.: MFCD00003424
Formel: Al₂O₃
MW: 101,96 g/mol
Kokpunkt: 2980 °C (1013 hPa)
Smältpunkt: 2045 °C
Densitet: 3,94 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003424
CAS nummer: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 13,00355