Aluminiumoxid substrate 10x10x0.5 mm, polished one side A plane
Leverantör: Thermo Scientific
Synonyms: Aluminium(III) oxide, Alumina C
38498.KT 38498.KU
38498.KTEA 2400 SEK
38498.KT 38498.KU
Aluminiumoxid substrate 10x10x0.5 mm, polished one side A plane
Aluminium oxide
Molecular Formula: Al2O3 Odour: Odourless MDL No.: MFCD00003424
Formel: Al₂O₃
MW: 101,96 g/mol
Kokpunkt: 2980 °C (1013 hPa)
Smältpunkt: 2045 °C
Densitet: 3,94 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003424
CAS nummer: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 13,00355