Magnesium sulphate hydrate, dried contains ca 1-2 mol water of hydration
Leverantör: Thermo Scientific
L13739.22 L13739.36 L13739.0E
L13739.22EA 520 SEK
L13739.22 L13739.36 L13739.0E
Magnesium sulphate hydrate, dried contains ca 1-2 mol water of hydration
Magnesium sulphate hydrate
CAS No.: 22189-08-8 Molecular Formula: MgSO4.xH2O Formula Weight: 120.37(anhy) Merck Reference: 14,5691 MDL No.: MFCD00149787
Formel: MgSO₄·nH₂O
MW: 120,37 g/mol (anhy)
Densitet: 2,45 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011110
CAS nummer: 22189-08-8
EINECS: 231-298-2
Merck Index: 14,5691