Kalciumsulfat, vattenfri ≥99.993%, Puratronic®
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Calcium sulphate (anhydrous) anhydrite, Gypsum
11116.04 11116.09 11116.18
11116.04EA 1300 SEK
11116.04 11116.09 11116.18
Kalciumsulfat, vattenfri ≥99.993%, Puratronic®
Kalciumsulfat
Purity: 99.993% CAS No.: 7778-18-9 Molecular Formula: CaO4S Formula Weight: 136.14 Physical Form: Powder Density: 2.960 Appearance: White Odour: Odourless Melting Point: 1450° Merck Reference: 14,1706 MDL No.: MFCD00010912
Formel: CaO₄S
MW: 136.14 g/mol
Smältpunkt: 1450 °C
Densitet: 2.960
Storage Temperature: Omgivande
MDL Number: MFCD00010912
CAS nummer: 7778-18-9
EINECS: 231-900-3
Merck Index: 14,1706