Kalciumsulfid ≥99.9% (max. 0.05% Sr)
Leverantör: Thermo Fisher Scientific
Varning
13121.06 13121.14
13121.06EA 1675 SEK
13121.06 13121.14
Kalciumsulfid ≥99.9% (max. 0.05% Sr)
Kalciumsulfid
Purity: 99.9% CAS No.: 20548-54-3 Molecular Formula: CaS Formula Weight: 72.14 Physical Form: -325 Mesh Powder Density: 2.5 Melting Point: dec. Merck Reference: 14,1707 UN No.: 3134 MDL No.: MFCD00015990
Formel: CaS
MW: 72.14 g/mol
Smältpunkt: Dec.
Densitet: 2.5
Storage Temperature: Omgivande
MDL Number: MFCD00015990
CAS nummer: 20548-54-3
EINECS: 243-873-5
UN: 3134
ADR: 4.3 III
Merck Index: 14,1707