Kalciumfosfat, vattenfri ≥98%
Leverantör: Thermo Fisher Scientific
Synonyms: Calcium phosphate dibasic, Calcium hydrogen phosphate, Calcium perphosphate, ammonified, Anhydrous Dibasic Calcium Phosphate, Calcium phosphate, dibasic
40232.30 40232.A1 40232.A7
40232.30EA 1625 SEK
40232.30 40232.A1 40232.A7
Kalciumfosfat, vattenfri ≥98%
Kalciumfosfat
Purity: 98% min CAS No.: 7757-93-9 Molecular Formula: CaHO4P Formula Weight: 136.06 Physical Form: Powder Appearance: White Odour: Odourless Merck Reference: 14,1692 MDL No.: MFCD00010909
Formel: CaHPO₄
MW: 136.06 g/mol
Smältpunkt: 370
Densitet: 2.89 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00010909
CAS nummer: 7757-93-9
EINECS: 231-826-1
Merck Index: 14,1692