Kalciumhydroxid ≥95.0% ACS
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Slaked lime, Calcium dihydroxide
14662.A1 14662.30
14662.A1EA 1600 SEK
14662.A1 14662.30
Kalciumhydroxid ≥95.0% ACS
Kalciumhydroxid
Purity: 95.0% min CAS No.: 1305-62-0 Molecular Formula: CaH2O2 Formula Weight: 74.09 Physical Form: Powder Density: 2.24 Appearance: White Odour: Odourless Solubility: Slightly soluble in water. Soluble in glycerol, acids. Insoluble in alcohol Merck Reference: 14,1673 UN No.: 3262 MDL No.: MFCD00010901
Mortar, plasters, cements, gas manufacturing to remove ammonia, in lubricants, water softening, petrochemicals, soil conditioner, paving materials
Formel: Ca(OH)₂
MW: 74.09 g/mol
Smältpunkt: 550
Densitet: 2.24
Storage Temperature: Omgivande
MDL Number: MFCD00010901
CAS nummer: 1305-62-0
EINECS: 215-137-3
UN: 3262
ADR: 8 III
Merck Index: 14,1673