Barium hydroxide, anhydrous 94-98
Leverantör: Thermo Fisher Scientific
Fara
12195.18 12195.30 12195.A1
12195.18EA 940 SEK
12195.18 12195.30 12195.A1
Barium hydroxide, anhydrous 94-98
Barium hydroxide
Purity: 94-98% CAS No.: 17194-00-2 Molecular Formula: Ba(OH)2 Formula Weight: 171.36 Physical Form: Powder Sensitivity: Air Sensitive and Hygroscopic Appearance: White Odour: Odourless pH: 12.5 Melting Point: 408° Merck Reference: 14,977 UN No.: 3262 MDL No.: MFCD00003443
Formel: H₂BaO₂
MW: 171.34 g/mol
Smältpunkt: 408 °C
Densitet: 2.18 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003443
CAS nummer: 17194-00-2
EINECS: 241-234-5
UN: 3262
ADR: 8 III
Merck Index: 14,977