Magnesiumoxid substrate 10x10x0.5mm, polished one side 100 orientation
Leverantör: Thermo Fisher Scientific
Synonyms: Magnesia usta
36427.KT 36427.KU
36427.KTEA 3200 SEK
36427.KT 36427.KU
Magnesiumoxid substrate 10x10x0.5mm, polished one side 100 orientation
Magnesium oxide
CAS No.: 1309-48-4 Molecular Formula: MgO MDL No.: MFCD00011109
Formel: MgO
MW: 40.3 g/mol
Kokpunkt: 3600
Smältpunkt: 2800
Densitet: 3.58 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011109
CAS nummer: 1309-48-4
EINECS: 215-171-9
Merck Index: 13,05700