Magnesiumoxid ≥99.95%
Leverantör: Thermo Scientific
Synonyms: Magnesia usta
14684.14 14684.22 14684.36
14684.14EA 3075 SEK
14684.14 14684.22 14684.36
Magnesiumoxid ≥99.95%
Magnesium oxide
Purity: 99.95% CAS No.: 1309-48-4 Molecular Formula: MgO Formula Weight: 40.30 Physical Form: -325 Mesh Powder Density: 3.58 Refractive Index: 1.736 Odour: Odourless Melting Point: 2852° Boiling Point: 3600° Merck Reference: 14,5677 MDL No.: MFCD00011109
Formel: MgO
MW: 40,3 g/mol
Kokpunkt: 3600 °C
Smältpunkt: 2852 °C
Densitet: 3,58
Storage Temperature: Omgivande
MDL Number: MFCD00011109
CAS nummer: 1309-48-4
EINECS: 215-171-9
Merck Index: 14,5677