Kalciumoxid ≥99.998% (excluding other alkaline earth and alkali metals 130 ppm max), Puratronic®
Leverantör: Thermo Scientific
Fara
Synonyms: Quicklime, Calcium oxide from marble, caustic, Lime
10684.04 10684.09 10684.18 10684.30 10684.03
10684.04EA 1500 SEK
10684.04 10684.09 10684.18 10684.30 10684.03
Kalciumoxid ≥99.998% (excluding other alkaline earth and alkali metals 130 ppm max), Puratronic®
Kalciumoxid
Purity: 99.998% CAS No.: 1305-78-8 Molecular Formula: CaO Formula Weight: 56.08 Physical Form: Powder Density: 3.37 Refractive Index: 1.8300 Sensitivity: Air and Moisture Sensitive Appearance: White Odour: Odourless Melting Point: 2572° Boiling Point: 2850° Merck Reference: 14,1686 MDL No.: MFCD00010911
Formel: CaO
MW: 56,08 g/mol
Kokpunkt: 2850 °C
Smältpunkt: 2572 °C
Densitet: 3,37
Storage Temperature: Omgivande
MDL Number: MFCD00010911
CAS nummer: 1305-78-8
EINECS: 215-138-9
UN: 1910
ADR: 8,III
Merck Index: 14,1686