Magnesium iodide, ultra dry ≥99.996%
Leverantör: Thermo Fisher Scientific
Ampuled under argon
14283.03 14283.09
14283.03EA 3500 SEK
14283.03 14283.09
Magnesium iodide, ultra dry ≥99.996%
Magnesium iodide
Purity: 99.996% CAS No.: 10377-58-9 Molecular Formula: I2Mg Formula Weight: 278.12 Physical Form: -10 Mesh Beads, Ampouled under argon Sensitivity: Light Sensitive and Hygroscopic Appearance: White Odour: Odourless Melting Point: 637° dec. Merck Reference: 14,5671 MDL No.: MFCD00049485
Formel: MgI₂
MW: 278.11 g/mol
Smältpunkt: 637 °C (dec.)
Densitet: 3.89 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00049485
CAS nummer: 10377-58-9
EINECS: 233-825-1
Merck Index: 14,5671