Kalciumkarbonat ≥99.0% ACS låg alkalihalt
Leverantör: Thermo Scientific
Synonyms: Marble
36337.30 36337.A1
36337.30EA 1375 SEK
36337.30 36337.A1
Kalciumkarbonat ≥99.0% ACS låg alkalihalt
Kalciumkarbonat
Purity: 99.0% min CAS No.: 471-34-1 Molecular Formula: CaCO3 Formula Weight: 100.09 Physical Form: Powder Density: 2.930 Refractive Index: 1.6583 Appearance: White Odour: Odourless Melting Point: 800° dec. Merck Reference: 14,1657 MDL No.: MFCD00010906
Formel: CaCO₃
MW: 100,09 g/mol
Smältpunkt: 800 °C (dec.)
Densitet: 2,930
Storage Temperature: Omgivande
MDL Number: MFCD00010906
CAS nummer: 471-34-1
EINECS: 207-439-9
Merck Index: 14,1657