Bariumkarbonat 99.0-101.0% ACS
Leverantör: Thermo Fisher Scientific
Varning
36236.36 36236.A1 36236.D3
36236.36EA 2050 SEK
36236.36 36236.A1 36236.D3
Bariumkarbonat 99.0-101.0% ACS
Bariumkarbonat
Purity: 99.0-101.0% CAS No.: 513-77-9 Molecular Formula: BaCO3 Formula Weight: 197.35 Physical Form: Powder Density: 4.43 Melting Point: 811° Boiling Point: dec. Merck Reference: 14,969 MDL No.: MFCD00003448
Formel: BaCO₃
Kokpunkt: Dec.
Smältpunkt: 811 °C
Densitet: 4,43
Storage Temperature: Omgivande
MDL Number: MFCD00003448
CAS nummer: 513-77-9
EINECS: 208-167-3
UN: 1564
ADR: 6.1,III
Merck Index: 14,969