Magnesiumperklorat, vattenfri ACS
Leverantör: Thermo Scientific
Fara
Synonyms: Anhydrone
11636.22 11636.36
11636.22EA 2050 SEK
11636.22 11636.36
Magnesiumperklorat, vattenfri ACS
Magnesiumperklorat
CAS No.: 10034-81-8 Molecular Formula: Mg(ClO4)2 Formula Weight: 223.23 Physical Form: Granular Density: 2.21 Sensitivity: Hygroscopic Merck Reference: 14,5678 UN No.: 1475 MDL No.: MFCD00011102
Desiccant. Drying agent for gases
Formel: Cl₂MgO₈
MW: 223,21 g/mol
Smältpunkt: 251 °C (dec.)
Densitet: 2,21
Storage Temperature: Omgivande
MDL Number: MFCD00011102
CAS nummer: 10034-81-8
EINECS: 233-108-3
UN: 1475
ADR: 5.1,II
Merck Index: 14,5678