Strontium bromide, ultra dry ≥99.995%
Leverantör: Thermo Fisher Scientific
Varning
Ampuled under argon
35717.03 35717.06
35717.03EA 2175 SEK
35717.03 35717.06
Strontium bromide, ultra dry ≥99.995%
Strontium bromide
Purity: 99.995% CAS No.: 10476-81-0 Molecular Formula: BrSr2 Formula Weight: 247.44 Physical Form: Powder, Ampouled under argon Density: 4.216 Appearance: White Odour: Odourless Melting Point: 643° Merck Reference: 14,8837 MDL No.: MFCD00049548
Formel: Br₂Sr
Kokpunkt: 2146 °C (1013 hPa)
Smältpunkt: 643 °C
Densitet: 4,216
Storage Temperature: Omgivande
MDL Number: MFCD00049548
CAS nummer: 10476-81-0
EINECS: 233-969-5
Merck Index: 14,8837