Strontium bromide, vattenfri ≥99% (max. 0.1% Ba)
Leverantör: Thermo Scientific
Varning
16791.18 16791.30
16791.18EA 4925 SEK
16791.18 16791.30
Strontium bromide, vattenfri ≥99% (max. 0.1% Ba)
Strontium bromide
Purity: 99% CAS No.: 10476-81-0 Molecular Formula: Br2Sr Formula Weight: 247.44 Physical Form: Granular Density: 4.216 Appearance: White Odour: Odourless Melting Point: 643° Merck Reference: 14,8837 MDL No.: MFCD00049548
Formel: Br₂Sr
MW: 247,43 g/mol
Kokpunkt: 2146 °C (1013 hPa)
Smältpunkt: 643 °C
Densitet: 4,216
Storage Temperature: Omgivande
MDL Number: MFCD00049548
CAS nummer: 10476-81-0
EINECS: 233-969-5
Merck Index: 14,8837