Magnesium diboride ≥99%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Magnesium boride
88149.22 88149.06 88149.14
88149.22EA 9350 SEK
88149.22 88149.06 88149.14
Magnesium diboride ≥99%
Magnesium diboride
Purity: 99% CAS No.: 12007-25-9 Molecular Formula: B2Mg Formula Weight: 45.93 Physical Form: -100 Mesh Powder Appearance: Dark grey Odour: Odourless Melting Point: 800° dec. MDL No.: MFCD00049752
Formel: B₂Mg
MW: 45.92 g/mol
Smältpunkt: 800 °C (dec.)
Densitet: 2.3 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00049752
CAS nummer: 12007-25-9
EINECS: 234-501-2