Rubidium sulphate ≥99.8%
Leverantör: Thermo Fisher Scientific
Varning
10567.09 10567.18
10567.09EA 1225 SEK
10567.09 10567.18
Rubidium sulphate ≥99.8%
Rubidium sulphate
Purity: 99.8% CAS No.: 7488-54-2 Molecular Formula: Rb2SO4 Formula Weight: 267.00 Physical Form: -20 Mesh Powder Density: 3.613 Refractive Index: 1.513 Odour: Odourless Melting Point: 1050° MDL No.: MFCD00011190
Formel: O₄Rb₂S
MW: 267 g/mol
Kokpunkt: 1708
Smältpunkt: 1050 °C
Densitet: 3.613
Storage Temperature: Omgivande
MDL Number: MFCD00011190
CAS nummer: 7488-54-2
EINECS: 231-301-7