Kaliumsulfat, pulver, EMPROVE® EXPERT BP, E515, Ph. Eur., JP, extra pure suitable for use as excipient, SAFC®
Leverantör: MERCK PRODUCTION CHEMICALS
1.05151.9029 1.05151.1000
1.05151.9029EA 0 SEK
1.05151.9029 1.05151.1000
Kaliumsulfat, pulver, EMPROVE® EXPERT BP, E515, Ph. Eur., JP, extra pure suitable for use as excipient, SAFC®
Potassium sulphate
Formel: K₂SO₄
Kokpunkt: 1698 °C (1013 hPa)
Smältpunkt: 1069 °C
Densitet: 2,66 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011388
CAS nummer: 7778-80-5
EINECS: 231-915-5
Merck Index: 13,07759