Litiumnitrat ≥99.5%, Reagent Grade
Leverantör: Honeywell Chemicals
Varning
227986-1KG 227986-25KG 227986-50KG
227986-1KGEA 2925 SEK
227986-1KG HONC227986-25KG HONC227986-50KG
Litiumnitrat ≥99.5%, Reagent Grade
Litiumnitrat
  • ≥95% for general laboratory use.
Formel: LiNO₃
MW: 68.95 g/mol
Kokpunkt: >450 °C (dec. below the boiling point)
Smältpunkt: 264 °C (1.013 hPa)
Densitet: 2.38 g/cm³
Storage Temperature: Omgivande
MDL Number: MFCD00011094
CAS nummer: 7790-69-4
EINECS: 232-218-9
UN: 2722
ADR: 5.1 III
Merck Index: 13,05558
Order Now
Specifikation testresultat
Identity (IR) complying
Assay of Li 9 - 11 %