Cesium fluoride ≥99%
Leverantör: Thermo Fisher Scientific
Fara
12885.14 12885.22
12885.14EA 1825 SEK
12885.14 12885.22
Cesium fluoride ≥99%
Cesium fluoride
Purity: 99% CAS No.: 13400-13-0 Molecular Formula: CsF Formula Weight: 151.90 Physical Form: Crystalline Density: 4.115 Appearance: White Odour: Odourless pH: 6.5-7.5 Melting Point: 682° Boiling Point: 1251° UN No.: 2923 MDL No.: MFCD00010960
Formel: CsF
MW: 151.9 g/mol
Kokpunkt: 1251 °C
Smältpunkt: 682 °C
Densitet: 4.115
Storage Temperature: Omgivande
MDL Number: MFCD00010960
CAS nummer: 13400-13-0
EINECS: 236-487-3
UN: 2923
ADR: 8 II