Natriumtetrafenylboratlösningen ≥99.5% ACS
Leverantör: Thermo Scientific
Fara
Synonyms: Tetraphenyl sodium borate, Sodium tetraphenylboron, Tetraphenylboron sodium salt
36430.04 36430.09 36430.18
36430.04EA 545 SEK
36430.04 36430.09 36430.18
Natriumtetrafenylboratlösningen ≥99.5% ACS
Natriumtetrafenylboratlösningen
Purity: 99.5% min CAS No.: 143-66-8 Molecular Formula: C24H20BNa Formula Weight: 342.23 Physical Form: Powder Sensitivity: Light Sensitive Appearance: White pH: 8 Melting Point: 300° Beilstein Reference: 3599783 Merck Reference: 14,8690 UN No.: 2811 MDL No.: MFCD00011494
Formel: (C₆H₅)₄BNa
MW: 342,22 g/mol
Smältpunkt: >300 °C
Densitet: 1,15 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011494
CAS nummer: 143-66-8
EINECS: 205-605-5
UN: 2811
ADR: 6.1,III
Merck Index: 14,8690