Salpetersyra 65%
Leverantör: Roth Carl
|
Fara
|
4989.1
4989.2
4989.3
ROTH4989.1EA
0
SEK
ROTH4989.1
ROTH4989.2
ROTH4989.3
Salpetersyra 65%
Salpetersyra
Formel:
HNO₃ Kokpunkt: 120,5 °C (1013 hPa) Smältpunkt: –42 °C Densitet: 1,4 g/cm³ (20 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00011349 CAS nummer: 7697-37-2 EINECS: 231-714-2 UN: 2031 ADR: 8,II Merck Index: 13,06608 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...