JOD-syra ≥99%
Leverantör: Thermo Scientific
Fara
A11925.0B A11925.18 A11925.22 A11925.30
A11925.0BEA 13375 SEK
A11925.0B A11925.18 A11925.22 A11925.30
JOD-syra ≥99%
JOD-syra
Purity: 99% CAS No.: 7782-68-5 Molecular Formula: HIO3 Formula Weight: 175.91 Odour: Odourless Melting Point: 110° dec. UN No.: 3085 MDL No.: MFCD00011348
Formel: HIO₃
MW: 175,91 g/mol
Smältpunkt: 110 °C (dec.)
Densitet: 4,629
Storage Temperature: Omgivande
MDL Number: MFCD00011348
CAS nummer: 7782-68-5
EINECS: 231-962-1
UN: 3085
ADR: 5.1,II
Merck Index: 14,5012