JOD-syra 99.6% ACS

Leverantör: Thermo Scientific
Fara

423811000 423815000
ACRO423811000EA 1475 SEK
ACRO423811000 ACRO423815000
JOD-syra 99.6% ACS
JOD-syra
Formel: HIO₃
MW: 175,91 g/mol
Smältpunkt: 110 °C
Densitet: 4,629 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011348
CAS nummer: 7782-68-5
EINECS: 231-962-1
UN: 3085
ADR: 5.1,II
Merck Index: 13,05034

Order Now

Specifikation testresultat

Appearance White to off-white Crystalline powder, crystals and chunks
Titration Iodometric ≥99.5 %
Residue after ignition ≤0.02 %
Chloride and Bromide ≤0.02 % (as Cl)
Iodide (I) passes test
Sulfate (SO4) ≤150 ppm
Iron (Fe) ≤20 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.01 %
Nitrogen compounds (as N) ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR