JOD-syra 99.5% for analysis
Leverantör: Thermo Scientific
Fara
206450500 206452500 206450010
ACRO206450500EA 920 SEK
ACRO206450500 ACRO206452500 ACRO206450010
JOD-syra 99.5% for analysis
JOD-syra
Formel: HIO₃
MW: 175,91 g/mol
Smältpunkt: 110 °C (dec.)
Densitet: 4,629 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011348
CAS nummer: 7782-68-5
EINECS: 231-962-1
UN: 3085
ADR: 5.1,II
Merck Index: 13,05034
Order Now
Specifikation testresultat
Appearance White to light beige Powder or crystals
Titration Iodometric ≥99.4 %
Iron (Fe) ≤30 ppm
Heavy metals (as Pb) ≤50 ppm