2,5-Dimethylfuran 99%
Leverantör: Thermo Scientific
Fara
176030250 176031000 176035000
ACRO176030250EA 905 SEK
ACRO176030250 ACRO176031000 ACRO176035000
2,5-Dimethylfuran 99%
2,5-Dimethylfuran
Formel: C₆H₈O
MW: 96,13 g/mol
Kokpunkt: 92…94 °C (760 mmHg)
Smältpunkt: –62 °C
Densitet: 0,9 g/cm³
Flampunkt: –1 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003250
CAS nummer: 625-86-5
EINECS: 210-914-3
UN: 1993
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear colorless to amber Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4400 to 1.4440 (20°C, 589 nm)
GC ≥98.5 %
Color scale ≤20 APHA
Titration RCOCl ≥99.4 %