5-(Hydroxymethyl)-2-furaldehyde ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: 5-Hydroxymethyl-2-furancarbaldehyde, 5-(hydroxymethyl)furan-2-carbaldehyde, 5-(Hydroxymethyl)furfural, 5-Hydroxymethylfurfural
A12475.03 A12475.06 A12475.14
A12475.03EA 1525 SEK
A12475.03 A12475.06 A12475.14
5-(Hydroxymethyl)-2-furaldehyde ≥97%
5-(Hydroxymethyl)-2-furaldehyde
Purity: 97% CAS No.: 67-47-0 Molecular Formula: C6H6O3 Formula Weight: 126.11 Storage Temperature: 2°C to 8°C Flash Point: 79°(174°F) Refractive Index: 1.5620 Melting Point: 32-35° Boiling Point: 114-116°/1mm Beilstein Reference: 110889 Merck Reference: 14,4832 MDL No.: MFCD00003234
Formel: C₆H₆O₃
MW: 126,11 g/mol
Kokpunkt: 114…116 °C (1 mmHg)
Smältpunkt: 32…35 °C
Densitet: 1,206
Flampunkt: 79 °C (174 °F)
Storage Temperature: Kylskåp
MDL Number: MFCD00003234
CAS nummer: 67-47-0
EINECS: 200-654-9
Merck Index: 14,4832