Fluoren 98+%
Leverantör: Thermo Scientific
Varning
Synonyms: 9H-Fluorene, Diphenylenmethane
156130250 156131000 156135000
ACRO156130250EA 490 SEK
ACRO156130250 ACRO156131000 ACRO156135000
Fluoren 98+%
Fluoren
Formel: C₁₃H₁₀
MW: 166,22 g/mol
Kokpunkt: 295 °C (1013 hPa)
Smältpunkt: 112…116 °C
Densitet: 1,2 g/cm³
Flampunkt: 151 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001111
CAS nummer: 86-73-7
EINECS: 201-695-5
UN: 3077
ADR: 9,III
Merck Index: 13,04181
Order Now
Specifikation testresultat
Appearance White to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 113 to 117 °C
Loss on drying ≤0.5 %
GC ≥98.0 %