9,10-Phenanthrenequinone 95%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Phenanthrene-9,10-dione
A11762.06 A11762.14 A11762.22
A11762.06EA 755 SEK
A11762.06 A11762.14 A11762.22
9,10-Phenanthrenequinone 95%
9,10-Phenanthrenequinone
Purity: 95% CAS No.: 84-11-7 Molecular Formula: C14H8O2 Formula Weight: 208.22 Flash Point: 245°(473°F) Melting Point: 207-212° Boiling Point: 360° Beilstein Reference: 608838 Merck Reference: 14,7213 UN No.: 3077 MDL No.: MFCD00001163
Formel: C₁₄H₈O₂
Kokpunkt: >360 °C
Smältpunkt: 207…212 °C
Densitet: 1,405
Flampunkt: 245 °C (473 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00001163
CAS nummer: 84-11-7
EINECS: 201-515-5
UN: 3077
ADR: 9,III
Merck Index: 14,7213