1,8-Dichloro-9,10-anthraquinone 96%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 1,8-Dichloroanthraquinone, 1,8-Dichloroanthracene-9,10-dione
B23421.14 B23421.22 B23421.36
B23421.14EA 1025 SEK
B23421.14 B23421.22 B23421.36
1,8-Dichloro-9,10-anthraquinone 96%
1,8-Dichloro-9,10-anthraquinone
Purity: 96% CAS No.: 82-43-9 Molecular Formula: C14H6Cl2O2 Formula Weight: 277.11 Melting Point: 201-203° MDL No.: MFCD00001191
Formel: C₁₄H₆Cl₂O₂
MW: 277.11 g/mol
Smältpunkt: 201…203 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001191
CAS nummer: 82-43-9
EINECS: 201-420-9