1-Methylpiperazine ≥98.0%

Leverantör: TCI EUROPE
Fara

M0553-25ML M0553-500ML M0553-100ML
TCIAM0553-25MLEA 230 SEK
TCIAM0553-25ML TCIAM0553-500ML TCIAM0553-100ML
1-Methylpiperazine ≥98.0%
1-Methylpiperazine
Formel: C₅H₁₂N₂
MW: 100,16 g/mol
Kokpunkt: 137°C
Smältpunkt: –6 °C
Densitet: 0,9085 g/cm³ (20 °C)
Flampunkt: 42°C
Storage Temperature: Omgivande
MDL Number: MFCD00005966
CAS nummer: 109-01-3
EINECS: 203-639-5
UN: 2924
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a6a8acf40824db0ae74d15c5399ab06910c7653&vid=3a6a8acf40824db0ae74d15c5399ab06910c7653&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR