Oktametylcyklotetrasiloxan ≥98%

Leverantör: Thermo Scientific
Varning

Synonyms: Volasil 244, 2,2,4,4,6,6,8,8-Octamethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane, Silicone fluid DC 244, Dow Corning 244® fluid

A15240.22 A15240.36
A15240.22EA 1350 SEK
A15240.22 A15240.36
Oktametylcyklotetrasiloxan ≥98%
Oktametylcyklotetrasiloxan

Purity: 98% CAS No.: 556-67-2 Molecular Formula: C8H24O4Si4 Formula Weight: 296.62 Flash Point: 56°(132°F) Refractive Index: 1.3960 Melting Point: 17-18° Boiling Point: 175-176° Beilstein Reference: 1787074 Merck Reference: 14,6747 UN No.: 1993 MDL No.: MFCD00003269

Formel: [-Si(CH₃)₂O-]₄
MW: 296,62 g/mol
Kokpunkt: 175…176 °C
Smältpunkt: 17…18 °C
Densitet: 0,956
Flampunkt: 56 °C (132 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00003269
CAS nummer: 556-67-2
EINECS: 209-136-7
UN: 1993
ADR: 3,III
Merck Index: 14,6747

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR