α-Tropolone ≥98%
Leverantör: Thermo Scientific
Synonyms: Tropolone, 2-Hydroxytropone, 2-Hydroxycyclohepta-2,4,6-trien-1-one, 1,2-Tropolone, 2-Hydroxycyclohepta-2,4,6-trienone, α-Tropolone
A11299.03 A11299.06 A11299.14
A11299.03EA 1025 SEK
A11299.03 A11299.06 A11299.14
α-Tropolone ≥98%
α-Tropolone
Purity: 98% CAS No.: 533-75-5 Molecular Formula: C7H6O2 Formula Weight: 122.12 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 50-55° Boiling Point: 115°/12mm Beilstein Reference: 1904978 MDL No.: MFCD00004158
Formel: C₇H₆O₂
MW: 122,12 g/mol
Kokpunkt: 115 °C (12 mmHg)
Smältpunkt: 50…55 °C
Flampunkt: >110 °C (230 °F)
Storage Temperature: Kylskåp
MDL Number: MFCD00004158
CAS nummer: 533-75-5
EINECS: 208-577-2