Tetramethyl-1,3-cyclobutanedione ≥99%
Leverantör: Thermo Scientific
Synonyms: Tetramethylcyclobutane-1,3-dione
A16297.09 A16297.18 A16297.30
A16297.09EA 1150 SEK
A16297.09 A16297.18 A16297.30
Tetramethyl-1,3-cyclobutanedione ≥99%
Tetramethyl-1,3-cyclobutanedione
Purity: 99% CAS No.: 933-52-8 Molecular Formula: C8H12O2 Formula Weight: 140.18 Melting Point: 112-115° Boiling Point: 120-122°/150mm MDL No.: MFCD00001331
Formel: C₈H₁₂O₂
MW: 140,18 g/mol
Kokpunkt: 120…122 °C (150 mmHg)
Smältpunkt: 112…115 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001331
CAS nummer: 933-52-8
EINECS: 213-269-6