Cyclopentyl isothiocyanate
Leverantör: Thermo Scientific
|
Fara
|
TL00485DA
TL00485EA
MAYBTL00485DAEA
0
SEK
MAYBTL00485DA
MAYBTL00485EA
Cyclopentyl isothiocyanate
Cyclopentyl isothiocyanate
Formel:
C₆H₉NS MW: 127.21 g/mol Kokpunkt: 95…100 °C (1013 hPa) Densitet: 1.03 g/cm³ (20 °C) |
MDL Number:
MFCD00040877 CAS nummer: 33522-03-1 UN: 2922 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...