Cyclopropanecarbonyl chloride 98%
Leverantör: Thermo Scientific
Fara
Synonyms: Cyclopropanecarboxylic chloride, Cyclopropanecarboxylic acid chloride
A11948.14 A11948.22
A11948.14EA 1100 SEK
A11948.14 A11948.22
Cyclopropanecarbonyl chloride 98%
Cyclopropanecarbonyl chloride
Purity: 98% CAS No.: 4023-34-1 Molecular Formula: C4H5ClO Formula Weight: 104.54 Flash Point: 23°(73°F) Refractive Index: 1.4520 Odour: Pungent Boiling Point: 119-120° Beilstein Reference: 471286 UN No.: 2920 MDL No.: MFCD00001277
Formel: C₄H₅ClO
MW: 104,54 g/mol
Kokpunkt: 119…120 °C
Densitet: 1,150
Flampunkt: 23 °C (73 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00001277
CAS nummer: 4023-34-1
EINECS: 223-684-4
UN: 2920
ADR: 8,II