Cyclohexanecarbonyl chloride ≥97%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Cyclohexanecarboxylic acid chloride, Cyclohexanecarboxylic chloride
A19824.14 A19824.22
A19824.14EA 805 SEK
A19824.14 A19824.22
Cyclohexanecarbonyl chloride ≥97%
Cyclohexanecarbonyl chloride
Purity: 97+% CAS No.: 2719-27-9 Molecular Formula: C7H11ClO Formula Weight: 146.62 Flash Point: 66°(150°F) Refractive Index: 1.4700 Boiling Point: 183-185° Beilstein Reference: 742163 UN No.: 3265 MDL No.: MFCD00001456
Formel: C₇H₁₁ClO
Kokpunkt: 183…185 °C
Smältpunkt: 29 °C
Densitet: 1,088
Flampunkt: 66 °C (150 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00001456
CAS nummer: 2719-27-9
EINECS: 220-322-7
UN: 3265
ADR: 8,II