Kumarin 99+%
Leverantör: Thermo Scientific
Varning
110530010 110530050 110532500
ACRO110530010EA 1375 SEK
ACRO110530010 ACRO110530050 ACRO110532500
Kumarin 99+%
Kumarin
Formel: C₉H₆O₂
Kokpunkt: 298 °C (1013 hPa)
Smältpunkt: 68…71 °C
Densitet: 0,935 g/cm³ (20 °C)
Flampunkt: 162 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006850
CAS nummer: 91-64-5
EINECS: 202-086-7
UN: 2811
ADR: 6.1,III
Merck Index: 13,02588
Order Now
Specifikation testresultat
Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 68 to 71 °C
GC ≥99.0 %
Specific gravity (20 °C) 0.868 to 0.888
Acid value ≤1 mg KOH/g