Bifenyl-2,2'-diol ≥99%
Leverantör: Thermo Scientific
Fara
Synonyms: 2,2'-Biphenol, 2,2-Biphenol, Biphenyl-2,2'-diol
A14800.14 A14800.22 A14800.36
A14800.14EA 505 SEK
A14800.14 A14800.22 A14800.36
Bifenyl-2,2'-diol ≥99%
Bifenyl-2,2'-diol
Purity: 99% CAS No.: 1806-29-7 Molecular Formula: C12H10O2 Formula Weight: 186.21 Flash Point: 154°(309°F) Melting Point: 107-111° Boiling Point: 314-316° Beilstein Reference: 1638363 MDL No.: MFCD00002210
Formel: C₁₂H₁₀O₂
MW: 186,21 g/mol
Kokpunkt: 314…316 °C
Smältpunkt: 107…111 °C
Densitet: 1,34
Flampunkt: 154 °C (309 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00002210
CAS nummer: 1806-29-7
EINECS: 217-303-0