Bifenyl-2-ol ≥99%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Hydroxibifenyl
A10592.0E A10592.30 A10592.36
A10592.0EEA 3625 SEK
A10592.0E A10592.30 A10592.36
Bifenyl-2-ol ≥99%
Bifenyl-2-ol
Purity: 99% CAS No.: 90-43-7 Molecular Formula: C12H10O Formula Weight: 170.21 Flash Point: 124°(255°F) Odour: Mild Boiling Point: 152-154°/15mm Beilstein Reference: 606907 Merck Reference: 14,7304 UN No.: 3077 MDL No.: MFCD00002208
Formel: C₁₂H₁₀O
MW: 170,21 g/mol
Kokpunkt: 152…154 °C (15 mmHg)
Smältpunkt: 54…58 °C
Densitet: 1,293
Flampunkt: 124 °C (255 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00002208
CAS nummer: 90-43-7
EINECS: 201-993-5
UN: 3077
ADR: 9,III
Merck Index: 14,7304