2-Bifenylamin 98%
Leverantör: Thermo Fisher Scientific
Varning
103220050 103220250
ACRO103220050EA 470 SEK
ACRO103220050 ACRO103220250
2-Bifenylamin 98%
2-Bifenylamin
Formel: C₁₂H₁₁N
Kokpunkt: 299 °C (1013 hPa)
Smältpunkt: 48…52 °C
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007701
CAS nummer: 90-41-5
EINECS: 201-990-9
Order Now
Specifikation testresultat
Appearance Brown to purple Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 48 to 52 °C
GC ≥97.5 %