Benzyl salicylate ≥99%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Benzyl 2-hydroxybenzoate
A16184.0E A16184.22 A16184.36
A16184.0EEA 4875 SEK
A16184.0E A16184.22 A16184.36
Benzyl salicylate ≥99%
Benzyl salicylate
Purity: 99% CAS No.: 118-58-1 Molecular Formula: C14H12O3 Formula Weight: 228.25 Flash Point: 137°(278°F) Refractive Index: 1.5804 Melting Point: 18-20° Boiling Point: 190°/14mm Beilstein Reference: 2115365 Merck Reference: 14,1144 MDL No.: MFCD00020034
Formel: C₁₄H₁₂O₃
Kokpunkt: 190 °C (14 mmHg)
Smältpunkt: 18…20 °C
Densitet: 1,170
Flampunkt: 137 °C (278 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00020034
CAS nummer: 118-58-1
EINECS: 204-262-9
Merck Index: 14,1144